missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NrCAM Antibody (S364-51), Novus Biologicals™
Mouse Monoclonal Antibody
£176.00 - £456.00
Specifications
| Antigen | NrCAM |
|---|---|
| Clone | S364-51 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence 1:100 |
| Applications | Western Blot, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620957
|
Novus Biologicals
NBP2-42191-0.025mg |
0.025 mg |
£176.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633919
|
Novus Biologicals
NBP2-42191 |
0.1 mg |
£456.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NrCAM Monoclonal antibody specifically detects NrCAM in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| NrCAM | |
| 1 mg/mL | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers | |
| PBS (pH 7.4), 50% Glycerol | |
| bravo, hBravo, KIAA0343Neuronal surface protein Bravo, MGC138845, MGC138846, neuronal cell adhesion molecule, Ng-CAM-related, NgCAM-related cell adhesion molecule, nr-CAM | |
| Fusion protein amino acids 30-845 (extracellular domain) of mouse NrCAM. Rat: 96% identity (795/822 amino acids identical). Human: 91% identity (753/822 amino acids identical) ∽50% identity with Neurofascin. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S364-51 | |
| Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Q810U4 | |
| 4897 | |
| IgG2a | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title