missing translation for 'onlineSavingsMsg'
Learn More

NRBP1 Antibody (4D2), Novus Biologicals™

Product Code. 18338599 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338599 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338599 Supplier Novus Biologicals Supplier No. H00029959M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NRBP1 Monoclonal antibody specifically detects NRBP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRBP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 4D2
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10-1:2000, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH01221
Gene Alias BCON3NRBPMADM, FLJ27109, FLJ35541, MUDPNP, multiple domain putative nuclear protein, myeloid leukemia factor 1 adaptor molecule, nuclear receptor binding protein, nuclear receptor binding protein 1, nuclear receptor-binding protein
Host Species Mouse
Immunogen NRBP1 (AAH01221, 436 a.a. ~ 535 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PAEVETRKVVLMQCNIESVEEGVKHHLTLLLKLEDKLNRHLSCDLMPNENIPELAAELVQLGFISEADQSRLTSLLEETLNKFNFARNSTLNSAAVTVSS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 29959
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.