missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NRBF2 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, Raji. Predicted reactivity: Mouse (84%), Rat (87%), Chicken (81%), Rhesus Monkey (98%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NRBF2 (nuclear receptor-binding factor 2) modulates transcriptional activation by target nuclear receptors. It is involved in starvation-induced autophagy probably by its association with PI3K complex I (PI3KC3-C1). NRBF2 stabilizes the PI3KC3-C1 assembly and enhances ATG14-linked lipid kinase activity of PIK3C3.
Specifications
Specifications
| Antigen | NRBF2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.77 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Nrbf2 |
| Gene Accession No. | Q96F24 |
| Gene Alias | Comodulator of PPAR and RXR; comodulator of PPAR and RXR 1; comodulator of PPAR and RXR 2; COPR; COPR1; COPR2; Nrbf2; NRBF-2; nuclear receptor binding factor 2; nuclear receptor binding factor-2; nuclear receptor-binding factor 2 |
| Gene Symbols | Nrbf2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?