missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRBF2 Antibody (OTI1A2), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73054C
This item is not returnable.
View return policy
Description
NRBF2 Monoclonal specifically detects NRBF2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| NRBF2 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NRBF2 | |
| Full length recombinant protein of human NRBF2 (NP_110386) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot | |
| OTI1A2 | |
| Western Blot | |
| Comodulator of PPAR and RXR, COPR, COPR1, COPR2, DKFZp564C1664, FLJ30395, NRBF-2, nuclear receptor binding factor 2, nuclear receptor binding factor-2, nuclear receptor-binding factor 2 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 29982.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction