missing translation for 'onlineSavingsMsg'
Learn More

NRBF2 Antibody (OTI1A2), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18409913 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409913 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409913 Supplier Novus Biologicals Supplier No. NBP273054AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NRBF2 Monoclonal antibody specifically detects NRBF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRBF2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1A2
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias Comodulator of PPAR and RXR, COPR, COPR1, COPR2, DKFZp564C1664, FLJ30395, NRBF-2, nuclear receptor binding factor 2, nuclear receptor binding factor-2, nuclear receptor-binding factor 2
Host Species Mouse
Immunogen Full length recombinant protein of human NRBF2 (NP_110386) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29982
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.