missing translation for 'onlineSavingsMsg'
Learn More

NRBF2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £549.00

Specifications

Antigen NRBF2
Dilution Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30226942
View Documents
Novus Biologicals
NBP3-38273-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30226584
View Documents
Novus Biologicals
NBP3-38273-100ul
100 μL
£549.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NRBF2 Polyclonal antibody specifically detects NRBF2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications

Specifications

NRBF2
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Unconjugated
Rabbit
Signal Transduction
PBS (pH 7.3), 50% glycerol
29982
IgG
Affinity purified
Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Purified
RUO
Human, Mouse, Rat
Comodulator of PPAR and RXR, COPR, COPR1, COPR2, DKFZp564C1664, FLJ30395, NRBF-2, nuclear receptor binding factor 2, nuclear receptor binding factor-2, nuclear receptor-binding factor 2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-287 of human NRBF2 (NP_110386.2).,, Sequence:, MEVMEGPLNLAHQQSRRADRLLAAGKYEEAISCHKKAAAYLSEAMKLTQSEQAHLSLELQRDSHMKQLLLIQERWKRAQREERLKAQQNTDKDAAAHLQTSHKPSAEDAEGQSPLSQKYSPSTEKCLPEIQGIFDRDPDTLLYLLQQKSEPAEPCIGSKAPKDDKTIIEEQATKIADLKRHVEFLVAENERLRKENKQLKAEKARLLKGPIEKELDVDADFVETSELWSLPPHAETATASSTWQKFAANTGKAKDIPIPNLPPLDFPSPELPLMELSEDILKGFMNN
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.