missing translation for 'onlineSavingsMsg'
Learn More

NRAS Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18433245 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433245 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433245 Supplier Novus Biologicals Supplier No. NBP281861AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NRAS Polyclonal antibody specifically detects NRAS in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRAS
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias ALPS4, GTPase NRas, HRAS1, neuroblastoma RAS viral (v-ras) oncogene homolog, N-ras, N-ras protein part 4, NRAS1, NS6, Transforming protein N-Ras, v-ras neuroblastoma RAS viral oncogene homolog
Host Species Rabbit
Immunogen NRAS antibody was raised against a 15 amino acid peptide near the carboxy terminus of human NRAS. The immunogen is located within the last 50 amino acids of NRAS. Amino Acid Squence: QYRMKKLNSSDDGTQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4893
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.