missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRARP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £364.00
Specifications
| Antigen | NRARP |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
NRARP Polyclonal antibody specifically detects NRARP in Mouse samples. It is validated for Western BlotSpecifications
| NRARP | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cardiovascular Biology, Cell Biology, Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 441478 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| MGC61598, NOTCH-regulated ankyrin repeat protein, notch-regulated ankyrin repeat-containing protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-114 of human NRARP (NP_001004354.1). MSQAELSTCSAPQTQRIFQEAVRKGNTQELQSLLQNMTNCEFNVNSFGPEGQTALHQSVIDGNLELVKLLVKFGADIRLANRDGWSALHIAAFGGHQDIVLYLITKAKYAASGR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title