Learn More
Invitrogen™ NRAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588772
Description
Immunogen sequence: MNVQPCSRCG YGVYPAEKIS CIDQIWHKAC FHCEVCKMML SVNNFVSHQK KPYCHAHNPK NNTFTSVYHT PLNLNVRTFP EAISGIHDQE DGEQCKSVFH WDMKSKDKEG APNRQPLANE RAYWTGYGEG NAWCPGALPD PEIVRMVEAR KSLGEEYTED YEQPRGKGSF PAMITPAYQR AKKANQLASQ VEYKRGHDER ISRFSTVVDT PELLRSKAGA QLQSDVRYTE DYEQQRGKGS FPAMITPAYQ IAKRANELAS DVRYHQQYQK EMRGMAGPAI GAEGILTREC ADQYGQGYPE; Positive Samples: SH-SY5Y.
May be involved in anchoring the terminal actin filaments in the myofibril to the membrane and in transmitting tension from the myofibrils to the extracellular matrix.
Specifications
| NRAP | |
| Polyclonal | |
| Unconjugated | |
| NRAP | |
| nebulin related anchoring protein; nebulin-related anchoring protein; nebulin-related-anchoring protein; NRAP; N-RAP | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4892 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q86VF7 | |
| NRAP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human NRAP (NP_0012483921). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.