missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NRAP Monoclonal Antibody (1E9)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524402
This item is not returnable.
View return policy
Description
Peptide Sequence: LRHAQKAHQL QSDVKYKSDL NLTRGVGWTP PGSYKVEMAR RAAELANARG LGLQGAYRGA EAVEAGDHQS GEVNPDATEI LHVKKKKA.
May be involved in anchoring the terminal actin filaments in the myofibril to the membrane and in transmitting tension from the myofibrils to the extracellular matrix.
Specifications
| NRAP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q86VF7 | |
| NRAP | |
| NRAP (NP_932326.2, 1640 a.a. ~ 1727 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 1E9 | |
| Unconjugated | |
| NRAP | |
| nebulin related anchoring protein; nebulin-related anchoring protein; nebulin-related-anchoring protein; NRAP; N-RAP | |
| Mouse | |
| Protein A | |
| RUO | |
| 4892 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction