missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NRAMP2/SLC11A2/DMT1 Monoclonal antibody specifically detects NRAMP2/SLC11A2/DMT1 in Human, Mouse, Porcine, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | NRAMP2/SLC11A2/DMT1 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 4C6 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500 |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_000608 |
| Gene Alias | DCT1NRAMP 2, Divalent cation transporter 1, Divalent metal transporter 1, DMT-1, DMT1FLJ37416, member 2, NRAMP2natural resistance-associated macrophage protein 2, solute carrier family 11 (proton-coupled divalent metal ion transporters) |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?