Learn More
Novus Biologicals NRAMP2/SLC11A2/DMT1 Antibody (2F7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals H00004891-M06
Product Code. 18309538
  • £453.00 / 0.01mg
Description
NRAMP2/SLC11A2/DMT1 Monoclonal antibody specifically detects NRAMP2/SLC11A2/DMT1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NRAMP2/SLC11A2/DMT1
Monoclonal
Unconjugated
In 1x PBS, pH 7.4
DCT1NRAMP 2, Divalent cation transporter 1, Divalent metal transporter 1, DMT-1, DMT1FLJ37416, member 2, NRAMP2natural resistance-associated macrophage protein 2, solute carrier family 11 (proton-coupled divalent metal ion transporters)
SLC11A2 (NP_000608, 1 a.a. ~ 65 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MVLGPEQKMSDDSVSGDHGESASLGNINPAYSNPSLSQSPGDSEEYFATYFNEKISIPEEEYSCF
0.1 mg
Primary
Human
Purified
Western Blot, ELISA, Sandwich ELISA
2F7
Western Blot 1:500, ELISA, Sandwich ELISA
NP_000608
Mouse
IgG purified
RUO
4891
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG2a κ