missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRAMP1/SLC11A1 Antibody (910825) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8400P
This item is not returnable.
View return policy
Description
NRAMP1/SLC11A1 Monoclonal antibody specifically detects NRAMP1/SLC11A1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| NRAMP1/SLC11A1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6556 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, CyTOF | |
| 910825 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| natural resistance-associated macrophage protein 1, LSH, member 1, NRAMP 1, NRAMP1solute carrier family 11 (sodium/phosphate symporters), member 1, NRAMPnatural resistance-associated macrophage protein 1, solute carrier family 11 (proton-coupled divalent metal ion transporters) | |
| NS0 mouse myeloma cell line transfected with human NRAMP1/SLC11A1, Met1-Gly550, Accession # NP_000569 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction