missing translation for 'onlineSavingsMsg'
Learn More

NRAGE/MAGED1 Antibody (353124) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30170386 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30170386 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30170386 Supplier Novus Biologicals Supplier No. FAB3835JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NRAGE/MAGED1 Monoclonal antibody specifically detects NRAGE/MAGED1 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRAGE/MAGED1
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 353124
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias DLXIN-1, MAGE tumor antigen CCF, melanoma antigen family D, 1, melanoma-associated antigen D1, Neurotrophin receptor-interacting MAGE homolog, NRAGE, NRAGE MAGE-D1 antigen
Host Species Mouse
Immunogen E. coli-derived recombinant human NRAGE, Met1-Glu778, Accession # Q9Y5V3
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 9500
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.