missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NR5A1 Recombinant Rabbit Monoclonal Antibody (3G7Z3)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537904
This item is not returnable.
View return policy
Description
Positive Samples: BxPC-3, Mouse ovary, Mouse spleen Immunogen sequence: AQELVLQLLA LQLDRQEFVC LKFIILFSLD LKFLNNHILV KDAQEKANAA LLDYTLCHYP HCGDKFQQLL LCLVEVRALS MQAKEYLYHK HLGNEMPRNN LLIEMLQAKQ T.
Steroidogenic factor-1 (SF-1) is an orphan nuclear receptor involved in organ development and differentiation of the endocrine system. SF-1 has been shown to be essential for fetal development of the hypothalamus, pituitary gland, gonads, and the adrenal gland. SF-1 has been shown to promote transcriptional activity of many steroidogenic enzymes from the cytochrome P450 family. SF-1 contains two activation function domains, designated AF1 and AF2, common to many nuclear steroid receptors which have been shown to be necessary for interaction with other nuclear receptors as well as coactivators and corepressors including steroid receptor coactivator-1 (SRC-1), glucocorticoid receptor interacting protein 1 (GRIP1), cAMP response element binding protein (CREB) binding protein (CBP) and silencing mediator of retinoid acid and thyroid hormone receptor (SMRT).
Specifications
| NR5A1 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P33242, P50569, Q13285 | |
| Nr5a1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 351-461 of human Steroidogenic factor-1 (SF-1/NR5A1) (Q13285). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 3G7Z3 | |
| Unconjugated | |
| Nr5a1 | |
| Ad4BP; Ad4BP/SF-1; adrenal 4 binding protein; Adrenal 4 binding protein (AD4BP); adrenal 4-binding protein; ELP; ELP-3; Embryonal long terminal repeat-binding protein; embryonal LTR-binding protein; FTZ1; FTZF1; Ftz-F1; fushi tarazu 1 factor homolog; fushi tarazu factor homolog 1; Fushi tarazu factor homolog 1 (FTZ1); hSF-1; Nr5a1; nuclear receptor AdBP4; nuclear receptor subfamily 5 group A member 1; Nuclear receptor subfamily 5 group A member 1 (NR5A1); nuclear receptor subfamily 5, group A, member 1; POF7; SF1; SF-1; SPGF8; SRXY3; Steroid hormone receptor Ad4BP; steroid hydroxylase positive regulator; Steroidogenic factor 1; Steroidogenic factor 1 (SF1); steroidogenic factor 1 nuclear receptor; steroidogenic factor-1; STF1; STF-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2516, 26423, 83826 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction