Learn More
NR4A3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55405-1-AP-150UL
This item is not returnable.
View return policy
Description
The NR4A3 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human NR4A3. This antibody recognizes human, mouse antigen. The NR4A3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| NR4A3 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 8013 | |
| IgG |
| Western Blot, ELISA | |
| 260 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |