missing translation for 'onlineSavingsMsg'
Learn More

NR2F6 Mouse anti-Human, Clone: 2H2B8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60117-2-IG-20UL

 View more versions of this product

Product Code. 16827085

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Transcription factor predomitly involved in transcriptional repression. Binds to promoter/enhancer response elements that contain the imperfect 5′-AGGTCA-3′ direct or inverted repeats with various spacings which are also recognized by other nuclear hormone receptors. Involved in modulation of hormonal responses. Represses transcriptional activity of the lutropin-choriogonadotropic hormone receptor/LHCGR gene, the renin/REN gene and the oxytocin-neurophysin/OXT gene. Represses the triiodothyronine-dependent and -independent transcriptional activity of the thyroid hormone receptor gene in a cell type-specific manner. The corepressing function towards thyroid hormone receptor beta/THRB involves at least in part the inhibition of THRB binding to triiodothyronine response elements (TREs) by NR2F6. Inhibits NFATC transcription factor DNA binding and subsequently its transcriptional activity. Acts as transcriptional repressor of IL-17 expression in Th-17 differentiated CD4(+) T cells and may be involved in induction and/or maintece of peripheral immunological tolerance and autoimmunity. Involved in development of forebrain circadian clock; is required early in the development of the locus coeruleus (LC).
Specifications

Specifications

NR2F6
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P10588
NR2F6
NR2F6 Fusion Protein Ag7614
20 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
2H2B8
Unconjugated
NR2F6
EAR 2, EAR2, ERBAL2, NR2F6, V erbA related protein 2
Mouse
Protein G
RUO
2063
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.