missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NR2F2 Recombinant Rabbit Monoclonal Antibody (K01_2N62)

Product Code. 30830066
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830066 100 μL
30826617 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830066 Supplier Invitrogen™ Supplier No. MA564146
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

NR2F2 Recombinant Monoclonal Antibody for IHC (P), Western Blot

This gene encodes a member of the steroid thyroid hormone superfamily of nuclear receptors. The encoded protein is a ligand inducible transcription factor that is involved in the regulation of many different genes. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR2F2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2N62
Concentration 0.80 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene NR2F2
Gene Accession No. P24468
Gene Alias 2700033K02Rik; 9430015G03Rik; ADP-ribosylation factor related protein 1; apolipoprotein A1 regulatory protein 1; apolipoprotein AI regulatory protein 1; apolipoprotein A-I regulatory protein 1; apolipoprotein regulatory protein 1; Aporp1; ARP1; ARP-1; chicken ovalbumin; chicken ovalbumin upstream promoter transcription factor 2; chicken ovalbumin upstream promoter-transcription factor I; CHTD4; COUP transcription factor 2; COUP transcription factor II; COUPb; COUP-TF II; COUP-TF2; COUPTFB; COUPTFbeta; COUPTFII; COUP-TFII; COUP-TFII transcription factor; Drosophila seven-up homolog/mammalian ARP-1 homolog; EAR3; MGC117452; NF-E3; NR2F1; NR2F2; Nuclear receptor subfamily 2 group F member 2; nuclear receptor subfamily 2, group F, member 1; nuclear receptor subfamily 2, group F, member 2; ovalbumin upstream promoter beta nuclear receptor; ovalbumin upstream promoter transcription factor II; seven-up related 40; svp[40]; SVP40; svp-related nuclear receptor/retinoid signaling modulator; Tcfcoup2; TFCOUP2; transcription factor COUP 2; wu:fc94g08
Gene Symbols NR2F2
Host Species Rabbit
Immunogen A synthetic peptide of human NR2F2.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7026
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.