missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NR2F2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541613
This item is not returnable.
View return policy
Description
Peptide sequence: QDEVPGSQGS QASQAPPVPG PPPGAPHTPQ TPGQGGPAST PAQTAAGGQG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 79%.
This gene encodes a member of the steroid thyroid hormone superfamily of nuclear receptors. The encoded protein is a ligand inducible transcription factor that is involved in the regulation of many different genes. Alternate splicing results in multiple transcript variants.
Specifications
| NR2F2 | |
| Polyclonal | |
| Unconjugated | |
| NR2F2 | |
| 2700033K02Rik; 9430015G03Rik; ADP-ribosylation factor related protein 1; apolipoprotein A1 regulatory protein 1; apolipoprotein AI regulatory protein 1; apolipoprotein A-I regulatory protein 1; apolipoprotein regulatory protein 1; Aporp1; ARP1; ARP-1; chicken ovalbumin; chicken ovalbumin upstream promoter transcription factor 2; chicken ovalbumin upstream promoter-transcription factor I; CHTD4; COUP transcription factor 2; COUP transcription factor II; COUPb; COUP-TF II; COUP-TF2; COUPTFB; COUPTFbeta; COUPTFII; COUP-TFII; COUP-TFII transcription factor; Drosophila seven-up homolog/mammalian ARP-1 homolog; EAR3; MGC117452; NF-E3; NR2F1; NR2F2; Nuclear receptor subfamily 2 group F member 2; nuclear receptor subfamily 2, group F, member 1; nuclear receptor subfamily 2, group F, member 2; ovalbumin upstream promoter beta nuclear receptor; ovalbumin upstream promoter transcription factor II; seven-up related 40; svp[40]; SVP40; svp-related nuclear receptor/retinoid signaling modulator; Tcfcoup2; TFCOUP2; transcription factor COUP 2; wu:fc94g08 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7026 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P24468 | |
| NR2F2 | |
| Synthetic peptide directed towards the N-terminal of human NR2F2 (aa 12-61). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction