Learn More
Invitrogen™ NR2F1 Recombinant Rabbit Monoclonal Antibody (24GB430)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557134
 View more versions of this product
Product Code. 30275335
  • £126.00 / 20µL
Description
NR2F1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene is a nuclear hormone receptor and transcriptional regulator. The encoded protein acts as a homodimer and binds to 5'-AGGTCA-3' repeats. Defects in this gene are a cause of Bosch-Boonstra optic atrophy syndrome (BBOAS).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NR2F1
Recombinant Monoclonal
0.65 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P10589, Q60632
Nr2f1
A synthesized peptide derived from human COUP TF1 (aa 1-53).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB430
Unconjugated
Nr2f1
avian erythroblastic leukemia viral (v-erb-a) oncogene homolog-like 3; BBOAS; BBSOAS; chicken ovalbumin upstream promoter-transcription factor I; COUP transcription factor 1; COUP transcription factor I; COUP-TF I; COUP-TF1; COUPTFA; COUP-TFI; EAR3; EAR-3; Erbal3; Nr2f1; NR2F2; Nuclear receptor subfamily 2 group F member 1; nuclear receptor subfamily 2, group F, member 1; SVP44; Tcfcoup1; TFCOUP1; transcription factor COUP 1 (chicken ovalbumin upstream promoter 1, v-erb-a homolog-like 3); V-erbA-related protein 3
Rabbit
Affinity Chromatography
RUO
13865, 7025, 81808
-20°C