missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NR2F1 Polyclonal Antibody

Product Code. 12835693
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12835693 100 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12835693 Supplier Invitrogen™ Supplier No. PA521611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: COUP TF1 tansfected 293T cell. Predicted reactivity: Mouse (100%), Rat (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a nuclear hormone receptor and transcriptional regulator. The encoded protein acts as a homodimer and binds to 5'-AGGTCA-3' repeats. Defects in this gene are a cause of Bosch-Boonstra optic atrophy syndrome (BBOAS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR2F1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 10% glycerol and 0.01% thimerosal; pH 7
Gene Nr2f1
Gene Accession No. P10589, Q60632
Gene Alias avian erythroblastic leukemia viral (v-erb-a) oncogene homolog-like 3; BBOAS; BBSOAS; chicken ovalbumin upstream promoter-transcription factor I; COUP transcription factor 1; COUP transcription factor I; COUP-TF I; COUP-TF1; COUPTFA; COUP-TFI; EAR3; EAR-3; Erbal3; Nr2f1; NR2F2; Nuclear receptor subfamily 2 group F member 1; nuclear receptor subfamily 2, group F, member 1; SVP44; Tcfcoup1; TFCOUP1; transcription factor COUP 1 (chicken ovalbumin upstream promoter 1, v-erb-a homolog-like 3); V-erbA-related protein 3
Gene Symbols Nr2f1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human COUP TF1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13865, 7025
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.