missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NR2C2AP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565965
This item is not returnable.
View return policy
Description
Immunogen sequence: MTHSLVCPETV SRVSSVLNRN TRQFGKKHLF DQDEETCWNS DQGPSQWVTL EFPQLIRVSQ LQIQFQGGF Highest antigen sequence identity to the following orthologs - mouse 86%, rat 86%.
May act as a repressor of NR2C2-mediated transactivation by suppressing the binding between NR2C2/TR4 and the TR4-response element in target genes.
Specifications
| NR2C2AP | |
| Polyclonal | |
| Unconjugated | |
| NR2C2AP | |
| 2310073E15Rik; Nr2c2ap; nuclear receptor 2C2 associated protein; nuclear receptor 2C2-associated protein; repressor for TR4 transactivation; TR4 orphan receptor associated protein TRA16; TR4 orphan receptor-associated 16 kDa protein; TRA16 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 126382 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86WQ0 | |
| NR2C2AP | |
| Recombinant Human NR2C2AP. Recombinant protein control fragment (Product #RP-105019). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction