missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NR2C2 Recombinant Rabbit Monoclonal Antibody (24GB730)

Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | NR2C2 |
|---|---|
| Clone | 24GB730 |
| Concentration | 1.5 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275991
![]() |
Invitrogen™
MA557518 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275739
|
Invitrogen™
MA557519 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NR2C2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Members of the nuclear hormone receptor family, such as NR2C2, act as ligand-activated transcription factors. The proteins have an N-terminal transactivation domain, a central DNA-binding domain with 2 zinc fingers, and a ligand-binding domain at the C terminus. The activated receptor/ligand complex is translocated to the nucleus where it binds to hormone response elements of target genes.Specifications
| NR2C2 | |
| 1.5 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P49116, P49117, P55094 | |
| 22026, 50659, 7182 | |
| A synthesized peptide derived from human NR2C2 / TR4 (aa 500-555). | |
| Antibody |
| 24GB730 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| NR2C2 | |
| mKIAA4145; Mtr2r1; Nr2c2; Nuclear hormone receptor TR4; nuclear receptor subfamily 2 group C member 2; nuclear receptor subfamily 2, group C, member 2; nuclear receptor subfamily 2, group H, member 2; orphan nuclear receptor TAK1; Orphan nuclear receptor TR4; orphan receptor TR4; orphan receptor, TR4; Tak1; testicular nuclear receptor 4; testicular receptor 4; TR2R1; Tr4; TR4 orphan receptor; TR4-NS orphan receptor | |
| NR2C2 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title