missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR2C2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20981-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to NR2C2.
Members of the nuclear hormone receptor family, such as NR2C2, act as ligand-activated transcription factors. The proteins have an N-terminal transactivation domain, a central DNA-binding domain with 2 zinc fingers, and a ligand-binding domain at the C terminus. The activated receptor/ligand complex is translocated to the nucleus where it binds to hormone response elements of target genes.Specifications
| NR2C2 | |
| Polyclonal | |
| Unconjugated | |
| NR2C2 | |
| hTAK1, NR2C2, Orphan nuclear receptor TAK1, Orphan nuclear receptor TR4, TAK1, Testicular receptor 4, TR2R1, TR4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22026, 7182 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P49116, P49117 | |
| NR2C2 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction