missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NR2C2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528194
This item is not returnable.
View return policy
Description
Recommended positive controls: H1299, Molt-4. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (81%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Members of the nuclear hormone receptor family, such as NR2C2, act as ligand-activated transcription factors. The proteins have an N-terminal transactivation domain, a central DNA-binding domain with 2 zinc fingers, and a ligand-binding domain at the C terminus. The activated receptor/ligand complex is translocated to the nucleus where it binds to hormone response elements of target genes.
Specifications
| NR2C2 | |
| Polyclonal | |
| Unconjugated | |
| NR2C2 | |
| mKIAA4145; Mtr2r1; Nr2c2; Nuclear hormone receptor TR4; nuclear receptor subfamily 2 group C member 2; nuclear receptor subfamily 2, group C, member 2; nuclear receptor subfamily 2, group H, member 2; orphan nuclear receptor TAK1; Orphan nuclear receptor TR4; orphan receptor TR4; orphan receptor, TR4; Tak1; testicular nuclear receptor 4; testicular receptor 4; TR2R1; Tr4; TR4 orphan receptor; TR4-NS orphan receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7182 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.42 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P49116 | |
| NR2C2 | |
| Recombinant protein encompassing a sequence within the center region of human NR2C2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction