missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR1H4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25055-1-AP-150UL
This item is not returnable.
View return policy
Description
The Vasoactive Intestinal Polypeptide Receptor 2 (VPAC2) is a member of a family of three G-protein coupled receptors which mediate VIP (vasoactive intestinal peptide) and PACAP (pituitary adenlyate cyclase activating peptide) function in a variety of tissues. Together, the VPAC1 and VPAC2 receptors compose a subtype of the VIP receptor. Expression of the VIP and PACAP receptors has been linked to the presence of human tumors.Specifications
| NR1H4 | |
| Polyclonal | |
| Unconjugated | |
| NR1H4 | |
| BAR, Bile acid receptor, Farnesoid X activated receptor, Farnesol receptor HRR 1, FXR, HRR 1, HRR1, NR1H4, RIP14, RXR interacting protein 14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9971 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96RI1 | |
| NR1H4 | |
| NR1H4 Fusion Protein Ag21878 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction