missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NR1D1 Recombinant Rabbit Monoclonal Antibody (K01_1U89)

Product Code. 30829346
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829346 100 μL
30827318 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829346 Supplier Invitrogen™ Supplier No. MA563762

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: KLPDLRTLNN MHSEK.

This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR1D1
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1U89
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene NR1D1
Gene Accession No. P20393, Q3UV55, Q63503
Gene Alias A530070C09Rik; BD73; Ear1; ear-1; EAR-1R; HREV; NR1D1; nuclear receptor protein; nuclear receptor Rev-ErbA-alpha; nuclear receptor subfamily 1 group D member 1; nuclear receptor subfamily 1, group D, member 1; orphan nuclear receptor; R75201; rev-erb alpha; Rev-erba; rev-erbA alpha; rev-erbA(alpha); Reverbaalpha; Rev-ErbAalpha; rev-erbA-alpha; Rev-ErbA-alpha protein; REV-ERBalpha; RVR; THRA1; THRAL; V-erbA-related protein 1; V-erbA-related protein EAR-1
Gene Symbols NR1D1
Host Species Rabbit
Immunogen A synthetic peptide of human NR1D1 (aa 591-605).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217166, 252917, 9572
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.