missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NR1D1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533999
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset (100%) Mouse, Rat, Hamster, Panda, Dog, Bovine, Horse, Rabbit (94%).
This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.
Specifications
| NR1D1 | |
| Polyclonal | |
| Unconjugated | |
| NR1D1 | |
| A530070C09Rik; BD73; Ear1; ear-1; EAR-1R; HREV; NR1D1; nuclear receptor protein; nuclear receptor Rev-ErbA-alpha; nuclear receptor subfamily 1 group D member 1; nuclear receptor subfamily 1, group D, member 1; orphan nuclear receptor; R75201; rev-erb alpha; Rev-erba; rev-erbA alpha; rev-erbA(alpha); Reverbaalpha; Rev-ErbAalpha; rev-erbA-alpha; Rev-ErbA-alpha protein; REV-ERBalpha; RVR; THRA1; THRAL; V-erbA-related protein 1; V-erbA-related protein EAR-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9572 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P20393 | |
| NR1D1 | |
| Synthetic 18 amino acid peptide from internal region of human NR1D1. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction