missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NR12S
GREENER_CHOICE

Product Code. 18759686
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18759686

Brand: Tocris Bioscience™ 7509/100U

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorogenic lipid membrane dye

NR12S is a fluorogenic lipid membrane dye that stains the outer leaflet of lipid vesicles and cells. In living cells the color emission varies with cholesterol content. Excitation maximum = 554 nm; emission maximum = 627 nm in DMSO.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-((3-((9-(Diethylamino)-5-oxo-5 H-benzo[a]phenoxazin-2-yl)oxy)propyl)(dodecyl)(methyl)ammonio)propane-1-sulfonate
CAS 1221739-85-0
Quantity 100 μg
Target Fluorescent Lipids Probes and Cell Membrane Stains
Molecular Formula C39H57N3O6S
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.