missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531365
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (80%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes an NADPH-dependent diflavin reductase that contains both flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD) binding domains. The encoded protein is an enzyme that catalyzes the transfers electrons from NADPH through FAD and FMN cofactors to potential redox partners. Alternative splicing results in multiple transcript variants.
Specifications
| NR1 | |
| Polyclonal | |
| Unconjugated | |
| NDOR1 | |
| 4930447P04Rik; bA350O14.9; NADPH dependent diflavin oxidoreductase 1; NADPH-dependent diflavin oxidoreductase 1; NADPH-dependent FMN and FAD-containing oxidoreductase; Ndor; NDOR1; Novel reductase 1; NR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27158 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9UHB4 | |
| NDOR1 | |
| Recombinant protein encompassing a sequence within the center region of human NDOR1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction