missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NR 7h
Description
NR 7h is a potent and selective p38α and p38β Degrader (PROTAC™) (DC50 < 50 nM). Displays no significant degradation of p38γ, p38δ, JNK1/2 or ERK1/2. Inhibits phosphorylation of MK2 in UV-treated cancer cells and LPS-stimulated bone marrow-derived macrophages (BMDM). Exhibits similar effect to p38α gene knockout in BBL358 cells. NR 7h impairs replication of Mayaro virus in human dermal fibroblasts and HeLa cells. Active in vivo.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 3-(3-Bromo-4-((2,4-difluorobenzyl)oxy)-6-methyl-2-oxopyridin-1(2 H)-yl)-N-(4-(1-(4-((4-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)butyl)amino)-4-oxobutyl)-1 H-1,2,3-triazol-4-yl)butyl)-4-methylbenzamide |
| CAS | 2550399-06-7 |
| Quantity | 5 mg |
| Target | Active Degraders |
| Molecular Formula | C48H50BrF2N9O8 |
| Formula Weight | 3636 |
| Purity | 97% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?