Learn More
Invitrogen™ NQO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596324
Description
Immunogen sequence: MAGKKVLIVY AHQEPKSFNG SLKNVAVDEL SRQGCTVTVS DLYAMNLEPR ATDKDITGTL SNPEVFNYGV ETHEAYKQRS LASDITDEQK KVREADLVIF QFPLYWFSVP AILKGWMDRV LCQGFAFDIP GFYDSGLLQG KLALLSVTTG GTAEMYTKTG VNGDSRYFLW PLQHGTLHFC GFKVLAPQIS FAPEIASEEE RKGMVAAWSQ RLQTIWKEEP IPCTAHWHFG Q; Positive Samples: HeLa; Cellular Location: Cytoplasm.
NQO2 is a flavoprotein that catalyzes the 2-electron reduction of various quinones, redox dyes, and the vitamin K menadione. NQO2 predominantly uses dihydronicotinamide riboside (NRH) as the electron donor (Wu et al., 1997).
Specifications
| NQO2 | |
| Polyclonal | |
| Unconjugated | |
| NQO2 | |
| DHQV; DIA6; EC 1.10.99.2; MGC94180; NAD(P)H dehydrogenase, quinone 2; NAD(P)H menadione oxidoreductase 2, dioxin inducible; NAD(P)H menadione oxidoreductase-1, dioxin-inducible-2; NAD(P)H quinone dehydrogenase 2; NMO2; NMOR2; NQO 2; Nqo2; NRH dehydrogenase; NRH dehydrogenase [quinone] 2; NRH: quinone oxidoreductase; NRH:quinone oxidoreductase 2; N-ribosyldihydronicotinamide:quinone reductase 2; OTTHUMP00000015948; OTTHUMP00000015949; OTTHUMP00000015953; Ox 2; Ox2; QR2; Quinone; Quinone reductase 2; quinone ribosyldihydronicotinamide dehydrogenase; ribosyldihydronicotinamide; ribosyldihydronicotinamide dehydrogenase; ribosyldihydronicotinamide dehydrogenase [quinone]; ribosyldihydronicotinamide dehydrogenase quinone | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4835 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16083 | |
| NQO2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-231 of human NQO2 (NP_0008952). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.