missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NQO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576740
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
NQO2 is a flavoprotein that catalyzes the 2-electron reduction of various quinones, redox dyes, and the vitamin K menadione. NQO2 predominantly uses dihydronicotinamide riboside (NRH) as the electron donor (Wu et al., 1997).
Specifications
| NQO2 | |
| Polyclonal | |
| Unconjugated | |
| NQO2 | |
| DHQV; DIA6; EC 1.10.99.2; MGC94180; NAD(P)H dehydrogenase, quinone 2; NAD(P)H menadione oxidoreductase 2, dioxin inducible; NAD(P)H menadione oxidoreductase-1, dioxin-inducible-2; NAD(P)H quinone dehydrogenase 2; NMO2; NMOR2; NQO 2; Nqo2; NRH dehydrogenase; NRH dehydrogenase [quinone] 2; NRH: quinone oxidoreductase; NRH:quinone oxidoreductase 2; N-ribosyldihydronicotinamide:quinone reductase 2; OTTHUMP00000015948; OTTHUMP00000015949; OTTHUMP00000015953; Ox 2; Ox2; QR2; Quinone; Quinone reductase 2; quinone ribosyldihydronicotinamide dehydrogenase; ribosyldihydronicotinamide; ribosyldihydronicotinamide dehydrogenase; ribosyldihydronicotinamide dehydrogenase [quinone]; ribosyldihydronicotinamide dehydrogenase quinone | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18105, 291084, 4835 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P16083, Q6AY80, Q9JI75 | |
| NQO2 | |
| Recombinant full length Human NQO2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction