missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NQO-1 Rabbit anti-Human, Clone: 2, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | NQO-1 |
|---|---|
| Clone | 2 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18254846
|
Novus Biologicals
NBP2-90044-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18241876
|
Novus Biologicals
NBP2-90044-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NQO-1 Monoclonal specifically detects NQO-1 in Human samples. It is validated for Western Blot.Specifications
| NQO-1 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Azoreductase, DHQU, DIA4azoreductase, diaphorase (NADH/NADPH) (cytochrome b-5 reductase), dioxin-inducible 1, DTDEC 1.6.5.2, DT-diaphorase, Menadione reductase, NAD(P)H dehydrogenase [quinone] 1, NAD(P)H dehydrogenase, quinone 1, NAD(P)H:menadione oxidoreductase 1, NAD(P)H:Quinone acceptor oxidoreductase type 1, NAD(P)H:quinone oxidoreductase 1, NAD(P)H:quinone oxireductase, NMOR1diaphorase-4, Phylloquinone reductase, QR1NMORI, Quinone reductase 1 | |
| Recombinant Human NQO-1 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, Lipid and Metabolism, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 1728 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title