missing translation for 'onlineSavingsMsg'
Få mere at vide

NQO-1 Antibody [mFluor Violet 450 SE], Novus Biologicals™

Artikelnummer. 30733276 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
0.1 mL
Pakningsstørrelse:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30733276 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30733276 Leverandør Novus Biologicals Leverandørnr. AF7567MFV450

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Sheep Polyclonal Antibody

NQO-1 Polyclonal antibody specifically detects NQO-1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen NQO-1
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias Azoreductase, DHQU, DIA4azoreductase, diaphorase (NADH/NADPH) (cytochrome b-5 reductase), dioxin-inducible 1, DTDEC 1.6.5.2, DT-diaphorase, Menadione reductase, NAD(P)H dehydrogenase [quinone] 1, NAD(P)H dehydrogenase, quinone 1, NAD(P)H:menadione oxidoreductase 1, NAD(P)H:Quinone acceptor oxidoreductase type 1, NAD(P)H:quinone oxidoreductase 1, NAD(P)H:quinone oxireductase, NMOR1diaphorase-4, Phylloquinone reductase, QR1NMORI, Quinone reductase 1
Host Species Sheep
Immunogen E. coli-derived recombinant human NQO-1, Met1-Lys274, Accession # P15559
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1728
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.