missing translation for 'onlineSavingsMsg'
Learn More

NQ301, MedChemExpress

Product Code. 16472686
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472686 25 mg 25mg
16453248 10 mg 10mg
16429796 50 mg 50mg
16415977 5 mg 5mg
16414516 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16472686 Supplier Medchem Express Supplier No. HY101054
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NQ301 is an antithrombotic agent; inhibits collagen-challenged rabbit platelet aggregation with an IC50 of 10 mg/mL.

NQ301 is an antithrombotic agent

Specifications

Chemical Name or Material NQ301
CAS 130089-98-4
Color Red
Purity Grade Notes Research
Molecular Formula C18H12ClNO3
Quantity 25 mg
Solubility Information DMSO : ≥ 29 mg/mL (89.03 mM)
SMILES O=C1C(NC2=CC=C(C(C)=O)C=C2)=C(Cl)C(C3=C1C=CC=C3)=O
Molecular Weight (g/mol) 325.75
Formula Weight 325.75
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.