missing translation for 'onlineSavingsMsg'
Learn More

NPY5R Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18350876 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350876 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350876 Supplier Novus Biologicals Supplier No. NBP309980100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NPY5R Polyclonal specifically detects NPY5R in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen NPY5R
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias neuropeptide Y receptor type 5, neuropeptide Y receptor Y5, NPYR5NPY5-R, NPY-Y5 receptor, NPYY5-R, Y5 receptor
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N region of human NPY5R (NP_006165.1). Peptide sequence DEYYNKTLATENNTAATRNSDFPVWDDYKSSVDDLQYFLIGLYTFVSLLG
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 4889
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.