missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPY5R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106850
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NPY5R.
Receptor for neuropeptide Y and peptide YY. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity. Seems to be associated with food intake. Could be involved in feeding disorders.
Specifications
| NPY5R | |
| Polyclonal | |
| Unconjugated | |
| Npy5r | |
| HGNC:7958; Neuropeptide Y receptor type 5; neuropeptide Y receptor type 5-like protein; neuropeptide Y receptor Y5; Neuropeptide Y5 receptor; Npy5; NPY5 receptor; Npy5r; NPY5-R; NPYR5; NPYY5; NPY-Y5 receptor; NPYY5-R; Y5 receptor; Y5R | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18168, 25340, 4889 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70342, Q15761, Q63634 | |
| Npy5r | |
| A synthesized peptide derived from human NPY5R(Accession Q15761), corresponding to amino acid residues I221-L271. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction