missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPY1R Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577516
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Neuropeptide Y receptors are a class of G-protein coupled receptors which are activated by the closely related peptide horomones neuropeptide Y, peptide YY and pancreatic polypeptide. These receptors are involved in control of a diverse set of behavioral processes including appetite, circadian rhythme, and anxiety.
Specifications
| NPY1R | |
| Polyclonal | |
| Unconjugated | |
| NPY1R | |
| NPY1R | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29358, 4886 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
| 0.75 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P21555, P25929 | |
| Npy1r | |
| Peptide (C)RLKRRNNMMDKMRDNK, corresponding to amino acid residues 237-252 of human NPY1 R | |
| 50 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction