missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPY Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581627
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Neuropeptide Y.
This gene encodes a neuropeptide that is widely expressed in the central nervous system and influences many physiological processes, including cortical excitability, stress response, food intake, circadian rhythms, and cardiovascular function. The neuropeptide functions through G protein-coupled receptors to inhibit adenylyl cyclase, activate mitogen-activated protein kinase, regulate intracellular calcium levels, and activate potassium channels. A polymorphism in this gene resulting in a change of leucine 7 to proline in the signal peptide is associated with elevated cholesterol levels, higher alcohol consumption, and may be a risk factor for various metabolic and cardiovascular diseases.
Specifications
| NPY | |
| Polyclonal | |
| Unconjugated | |
| NPY | |
| 0710005A05Rik; C-flanking peptide of NPY; CPON; involved in appetite regulation; LOC100912228; Neuropeptide tyrosine; Neuropeptide Y; neuropeptide Y precursor; Neuropeptide-Y; Npy; NPY02; OTTHUMP00000201947; prepro-neuropeptide Y; proneuropeptide Y; pro-neuropeptide Y; pro-neuropeptide Y-like; PYY4; RATNPY; RATNPY02 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 4852 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| P01303 | |
| NPY | |
| A synthetic peptide corresponding to the C-terminus of the Human NPY/Pro-neuropeptide Y. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction