missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPW Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£328.00 - £513.00
Specifications
| Antigen | NPW |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18297234
|
Novus Biologicals
NBP2-57337 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18691719
|
Novus Biologicals
NBP2-57337-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NPW Polyclonal specifically detects NPW in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NPW | |
| Polyclonal | |
| Rabbit | |
| Human | |
| HPPL8, L8, L8C, Neuropeptide W, PPL8, PPNPW, Prepro-Neuropeptide W, Preproprotein L8 | |
| NPW | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 283869 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:REAPLLLPSWVQELWETRRRSSQAGIPVRAPRSPRAPEPALEPESLDFSGAGQRLRRDVSRPAVDPAANR | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title