Learn More
Invitrogen™ NPSR1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5100959
Product Code. 16697434
  • £448.00 / 100µL
Description
Antibody detects endogenous levels of total NPSR1.
May be active in signaling pathway in an autocrine or paracrine fashion in several tissues. Receptor for neuropeptide S, it may mediate its action, such as inhibitory effects, on cell growth. Involved in pathogenesis of asthma and other IgE-mediated diseases.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NPSR1
Polyclonal
Unconjugated
NPSR1
9330128H10Rik; ASRT2; G protein-coupled receptor 154; G protein-coupled receptor PGR14; Gpr154; GPRA; G-protein coupled receptor 154; G-protein coupled receptor for asthma susceptibility; G-protein coupled receptor PGR14; MVTR; Neuropeptide S receptor; neuropeptide S receptor 1; NPSR; Npsr1; Pgr14; RGD1564154; vasopressin receptor-related receptor 1; vasotocin receptor; VRR1
Rabbit
Affinity chromatography
RUO
300458, 319239, 387129
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P0C0L6, Q6W5P4, Q8BZP8
NPSR1
A synthesized peptide derived from human NPSR1(Accession Q6W5P4), corresponding to amino acid residues V233-F283.
100 μL
Primary
Human, Mouse, Rat