missing translation for 'onlineSavingsMsg'
Learn More

DBeQ, MedChemExpress

Product Code. 16439467
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16439467

Brand: Medchem Express HY15945

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DBeQ is a selective, potent, reversible, and ATP-competitive p97 inhibitor, with an IC50 value of 1.5 μM and 1.6 μM for p97(wt) and p97(C522A), respectively; DBeQ also inhibits Vps4 with an IC50 of 11.5 μM.

DBeQ is a selective, potent, reversible, and ATP-competitive p97 inhibitor, with an IC50 value of 1.5 μM and 1.6 μM for p97(wt) and p97(C522A), respectively

Specifications

Chemical Name or Material DBeQ
Physical Form Solid
CAS 177355-84-9
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H20N4
For Use With (Application) Cancer-programmed cell death
Synonym JRF 12
Solubility Information DMSO : 35.71 mg/mL (104.90 mM; Need ultrasonic)
SMILES C1(NCC2=CC=CC=C2)=NC(NCC3=CC=CC=C3)=C4C=CC=CC4=N1
Molecular Weight (g/mol) 340.42
Formula Weight 340.42
Percent Purity 98.05%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.