missing translation for 'onlineSavingsMsg'
Learn More

NPS-1034, MedChemExpress

Product Code. 16444356
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16444356

Brand: Medchem Express HY100509

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NPS-1034 is a dual inhibitor of AXL and MET with IC50s of 10.3 and 48 nM, respectively.

NPS-1034 is a dual inhibitor of AXL and MET with IC50s of 10.3 and 48 nM, respectively.

Specifications

Chemical Name or Material NPS-1034
CAS 1221713-92-3
Color White
Purity Grade Notes Research
Molecular Formula C31H23F2N5O3
Quantity 100 mg
Solubility Information DMSO : 34 mg/mL (61.65 mM; Need ultrasonic)
SMILES O=C(C1=C(C)N(C)N(C2=CC=C(F)C=C2)C1=O)NC3=CC=C(OC4=C5C(NC=C5C6=CC=CC=C6)=NC=C4)C(F)=C3
Molecular Weight (g/mol) 551.54
Formula Weight 551.54
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.