missing translation for 'onlineSavingsMsg'
Learn More

NPRL3 Antibody (4E6), Novus Biologicals™

Product Code. 18328228 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328228 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328228 Supplier Novus Biologicals Supplier No. H00008131M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NPRL3 Monoclonal antibody specifically detects NPRL3 in Human, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPRL3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4E6
Conjugate Unconjugated
Dilution Western Blot, ELISA
Formulation In 1x PBS, pH 7.4
Gene Alias -14 gene protein, Alpha-globin regulatory element-containing gene protein, C16orf35, CGTHBAchromosome 16 open reading frame 35, conserved gene telomeric to alpha globin cluster, MAREHS-40, nitrogen permease regulator 3-like protein, nitrogen permease regulator-like 3 (S. cerevisiae), NPR3, Protein CGTHBA, RMD11
Host Species Mouse
Immunogen C16orf35 (NP_036207, 470 a.a. ~ 568 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LLPSGDSPLNQRMTENLLASLSEHERAAILSVPAAQNPEDLRMFARLLHYFRGRHHLEEIMYNENTRRSQLLMLFDKFRSVLVVTTHEDPVIAVFQALL
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8131
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.