Learn More
Invitrogen™ NPRL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595774
Description
Immunogen sequence: MGSGCRIECI FFSEFHPTLG PKITYQVPED FISRELFDTV QVYIITKPEL QNKLITVTAM EKKLIGCPVC IEHKKYSRNA LLFNLGFVCD AQAKTCALEP IVKKLAGYLT TLELESSFVS MEESKQKLVP IMTILLEELN ASGRCTLPID ESNTIHLKVI EQRPDPPVAQ EYDVPVFTKD KEDFFNSQWD LTTQQILPYI DGFRHIQKIS AEADVELNLV; Positive Samples: DU145, U-937, 293T, Rat lung.
NPRL2, also known as TUSC4 (tumor suppressor candidate 4), is a 380 amino acid protein that contains a bipartite nuclear localization signal and a granulin protein-binding domain. It is highly expressed in skeletal muscle, followed by brain, liver and pancreas, with lower expression in lung, kidney, placenta and heart. NPRL2 is also expressed in most lung cancer cell lines and may be involved in tumor suppression. NPRL2 may play a role in mismatch repair, cell cycle checkpoint signaling and activation of apoptotic pathways. It may also enhance the therapeutic efficacy of chemotherapy drugs such as cis-platin by resensitizing patients resistant to cisplatin treatment. The gene encoding NPRL2 is conserved between species and is expressed as two isoforms due to alternative splicing events.
Specifications
| NPRL2 | |
| Polyclonal | |
| Unconjugated | |
| NPRL2 | |
| 2810446G01Rik; G21; G21 protein; GATOR complex protein NPRL2; Gene 21 protein; homologous to yeast nitrogen permease (candidate tumor suppressor); nitrogen permase homolog; nitrogen permease regulator 2-like protein; nitrogen permease regulator-like 2; nitrogen permease regulator-like 2 (S. cerevisiae); NPR 2L; NPR L2; NPR like 2; NPR2; NPR2 like; NPR2L; NPR2-like protein; NPR2-like, GATOR1 complex subunit; NPRL 2; Nprl2; Tumor suppressor candidate 4; TUSC 4; TUSC 4 protein; TUSC4; TUSC4 protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10641, 363138 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.15 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8WTW4 | |
| NPRL2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human NPRL2 (NP_0065363). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.