missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPRB/NPR2 Antibody (SR1075), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £351.00
Specifications
| Antigen | NPRB/NPR2 |
|---|---|
| Clone | SR1075 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663064
|
Novus Biologicals
NBP3-22358-100ul |
100 μg |
£351.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604065
|
Novus Biologicals
NBP3-22358-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NPRB/NPR2 Monoclonal antibody specifically detects NPRB/NPR2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
| NPRB/NPR2 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AMD1;AMDM;ANPb;ANPRB;Atrial natriuretic peptide receptor 2;ECDM;GCB;GC-B;GUC2B;GUCY2B;NPRB;NPRBi;SNSK | |
| A synthesized peptide derived from human NPRB/NPR2 (Uniprot #: P20594) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1075 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology, GPCR, Neuroscience, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 4882 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title