missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPRB/NPR2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93665-0.1ml
This item is not returnable.
View return policy
Description
NPRB/NPR2 Polyclonal antibody specifically detects NPRB/NPR2 in Human samples. It is validated for Western Blot
Specifications
| NPRB/NPR2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| AMD1;AMDM;ANPb;ANPRB;Atrial natriuretic peptide receptor 2;ECDM;GCB;GC-B;GUC2B;GUCY2B;NPRB;NPRBi;SNSK | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-320 of human NPRB/NPR2 (NP_003986.2). SAKNDHYRTLVRTGPSAPKLGEFVVTLHGHFNWTARAALLYLDARTDDRPHYFTIEGVFEALQGSNLSVQHQVYAREPGGPEQATHFIRANGRIVYICGPLEMLHEILLQAQRENLTNGDYVFFYLDVFGESLRAGPTRATGRPWQDNRTREQAQALREAFQTVLVITYREPPNPEYQEFQNRLLIRARED | |
| 0.1 mL | |
| Cardiovascular Biology, GPCR, Neuroscience, Signal Transduction | |
| 4882 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction