missing translation for 'onlineSavingsMsg'
Learn More

NPRB/NPR2 Antibody (2A6), Novus Biologicals™

Product Code. 18338108 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338108 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338108 Supplier Novus Biologicals Supplier No. H00004882M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NPRB/NPR2 Monoclonal antibody specifically detects NPRB/NPR2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPRB/NPR2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA, Sandwich ELISA Capture
Classification Monoclonal
Clone 2A6
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. CN841943
Gene Alias AMD1,AMDM,ANPb,ANPRB,Atrial natriuretic peptide receptor 2,ECDM,GCB,GC-B,GUC2B,GUCY2B,NPRB,NPRBi,SNSK
Host Species Mouse
Immunogen NPR2 (NP_003986.2, 131 a.a. ~ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AKNDHYRTLVRTGPSAPKLGEFVVTLHGHFNWTARAALLYLDARTDDRPHYFTIEGVFEALQGSNLSVQHQVYAREPGGPEQATHFIRANGRIVYICGPL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cardiovascular Biology, GPCR, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4882
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.