missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPRB/NPR2 Antibody (19C7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26459-50ul
This item is not returnable.
View return policy
Description
NPRB/NPR2 Monoclonal antibody specifically detects NPRB/NPR2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry
Specifications
| NPRB/NPR2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
| 19C7 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| AMD1,AMDM,ANPb,ANPRB,Atrial natriuretic peptide receptor 2,ECDM,GCB,GC-B,GUC2B,GUCY2B,NPRB,NPRBi,SNSK | |
| A synthesized peptide derived from Human NPRB/NPR2 [UniProt P20594] | |
| 50 μL | |
| Cardiovascular Biology, GPCR, Neuroscience, Signal Transduction | |
| 4882 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction